4-(Trimethylsilyl)aniline ≥95%

Supplier: Ambeed
Danger

A724997-1G A724997-5G
77893-442EA 208 USD
77893-442 77893-443
4-(Trimethylsilyl)aniline ≥95%
4-(Trimethylsilyl)aniline

Precaución: for Research Use Only

Formula: C₉H₁₅NSi
MDL Number: MFCD00999279
CAS Number: 17889-23-5
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR