3-(Bromomethyl)benzeneboronic acid, neopentyl glycol ester ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-(3-(Bromomethyl)phenyl)-5,5-dimethyl-1,3,2-dioxaborinane

A724918-1G A724918-250MG
77749-477EA 64 USD
77749-477 77749-478
3-(Bromomethyl)benzeneboronic acid, neopentyl glycol ester ≥98%
3-(Bromomethyl)benzeneboronic acid, neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BBrO₂
MW: 282.969 g/mol
MDL Number: MFCD04973735
CAS Number: 223799-25-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR