(2S)-2-Amino-3-(4-(2-amino-6-(2,2,2-trifluoro-1-(3'-fluoro-[1,1'-biphenyl]-4-yl)ethoxy)pyrimidin-4-yl)phenyl)propanoic acid ≥98%

Supplier: Ambeed
Warning

A724867-10MG A724867-1MG A724867-5MG
77598-840EA 62.53 USD
77598-840 77598-842 77598-844
(2S)-2-Amino-3-(4-(2-amino-6-(2,2,2-trifluoro-1-(3'-fluoro-[1,1'-biphenyl]-4-yl)ethoxy)pyrimidin-4-yl)phenyl)propanoic acid ≥98%
(2S)-2-Amino-3-(4-(2-amino-6-(2,2,2-trifluoro-1-(3'-fluoro-[1,1'-biphenyl]-4-yl)ethoxy)pyrimidin-4-yl)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₇H₂₂F₄N₄O₃
MDL Number: MFCD19053188
CAS Number: 945976-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR