2,6-Difluoro-N-(1-(4-hydroxy-2-(trifluoromethyl)benzyl)-1H-pyrazol-3-yl)benzamide ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
GSK-7975A
A724629-10MG
A724629-1MG
A724629-5MG
77598-828EA
21.53
USD
77598-828
77598-830
77598-832
2,6-Difluoro-N-(1-(4-hydroxy-2-(trifluoromethyl)benzyl)-1H-pyrazol-3-yl)benzamide ≥98%
2,6-Difluoro-N-(1-(4-hydroxy-2-(trifluoromethyl)benzyl)-1H-pyrazol-3-yl)benzamide
Precaución: for Research Use Only
Formula:
C₁₈H₁₂F₅N₃O₂ MW: 397.2988 g/mol |
MDL Number:
MFCD28411366 CAS Number: 1253186-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...