ONC212 ≥98%

Supplier: Ambeed
Danger

Synonyms: 7-Benzyl-4-(4-(trifluoromethyl)benzyl)-2,4,6,7,8,9-hexahydroimidazo[1,2-a]pyrido[3,4-e]pyrimidin-5(1H)-one

A724585-100MG A724585-50MG
77598-826EA 251.13 USD
77598-826 77622-590
ONC212 ≥98%
ONC212

Precaución: for Research Use Only

Formula: C₂₄H₂₃F₃N₄O
MW: 440.46 g/mol
MDL Number: MFCD31619281
CAS Number: 1807861-48-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR