1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-thioxo-3,4-dihydropyrimidin-2(1H)-one ≥97%

Supplier: Ambeed
Warning

Synonyms: 4-Thio-2’-deoxyuridine

A724422-100MG A724422-250MG
77747-435EA 57 USD
77747-435 77747-436
1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-thioxo-3,4-dihydropyrimidin-2(1H)-one ≥97%
1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-thioxo-3,4-dihydropyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄S
MW: 244.26 g/mol
MDL Number: MFCD00058457
CAS Number: 5580-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR