Asiaticoside ≥98%

Supplier: Ambeed
Warning

Synonyms: (1S,2R,4aS,6aS,6bR,8aR,9R,10R,11R,12aR,12bR,14bS)-(2S,3R,4S,5S,6R)-6-((((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl 10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylate

A724420-10MG A724420-50MG A724420-100MG A724420-1MG
77617-964EA 12.36 USD
77617-964 77749-475 77749-473 77749-474
Asiaticoside ≥98%
Asiaticoside

Precaución: for Research Use Only

Formula: C₄₈H₇₈O₁₉
MW: 959.12 g/mol
MDL Number: MFCD06642601
CAS Number: 16830-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR