Larazotide acetate ≥98%

Supplier: Ambeed
Warning

A724380-10MG A724380-1MG A724380-5MG
77598-814EA 34.85 USD
77598-814 77598-816 77598-818
Larazotide acetate ≥98%
Larazotide acetate

Precaución: for Research Use Only

Formula: C₃₄H₅₉N₉O₁₂
MDL Number: MFCD31630844
CAS Number: 881851-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR