Heparan sulfate >50.0 IU/mg

Supplier: Ambeed
Warning

A724029-10MG A724029-1MG A724029-5MG
77598-808EA 150.67 USD
77598-808 77598-810 77622-580
Heparan sulfate >50.0 IU/mg
Heparan sulfate

Precaución: for Research Use Only

Formula: C₂₆H₃₆N₃O₃₉S₆
CAS Number: 9050-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR