(2-(2,2-Diphenylethoxy)acetyl)-L-arginine 2,2,2-trifluoroacetate ≥99%

Supplier: Ambeed
Warning

Synonyms: SB290157 trifluoroacetate

A723911-10MG A723911-1MG A723911-5MG
77598-802EA 79.95 USD
77598-802 77598-804 77598-806
(2-(2,2-Diphenylethoxy)acetyl)-L-arginine 2,2,2-trifluoroacetate ≥99%
(2-(2,2-Diphenylethoxy)acetyl)-L-arginine 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₂₄H₂₉F₃N₄O₆
MW: 526.51 g/mol
MDL Number: MFCD04974198
CAS Number: 1140525-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR