(2R,3S)-3-((6-(((4,6-Dimethylpyridin-3-yl)methyl)amino)-9-isopropyl-9H-purin-2-yl)amino)pentan-2-ol ≥98%

Supplier: Ambeed
Warning

A723815-50MG A723815-100MG A723815-250MG
77783-768EA 336 USD
77783-768 77783-766 77783-767
(2R,3S)-3-((6-(((4,6-Dimethylpyridin-3-yl)methyl)amino)-9-isopropyl-9H-purin-2-yl)amino)pentan-2-ol ≥98%
(2R,3S)-3-((6-(((4,6-Dimethylpyridin-3-yl)methyl)amino)-9-isopropyl-9H-purin-2-yl)amino)pentan-2-ol

Precaución: for Research Use Only

Formula: C₂₁H₃₁N₇O
MDL Number: MFCD31693062
CAS Number: 1070790-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR