3-(2-Chloro-6-methylphenyl)-7-((2-methoxy-4-(1-methylpiperidin-4-yl)phenyl)amino)-1-(5-methoxypyridin-2-yl)-3,4-dihydropyrimido[4,5-d]pyrimidin-2(1H)-one ≥98%

Supplier: Ambeed
Warning

Synonyms: YKL-05-099

A723778-10MG A723778-1MG A723778-5MG
77598-790EA 267.53 USD
77598-790 77598-792 77598-794
3-(2-Chloro-6-methylphenyl)-7-((2-methoxy-4-(1-methylpiperidin-4-yl)phenyl)amino)-1-(5-methoxypyridin-2-yl)-3,4-dihydropyrimido[4,5-d]pyrimidin-2(1H)-one ≥98%
3-(2-Chloro-6-methylphenyl)-7-((2-methoxy-4-(1-methylpiperidin-4-yl)phenyl)amino)-1-(5-methoxypyridin-2-yl)-3,4-dihydropyrimido[4,5-d]pyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₃₂H₃₄ClN₇O₃
MW: 600.11 g/mol
MDL Number: MFCD31382123
CAS Number: 1936529-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR