SKQ1 Bromide ≥98%

Supplier: Ambeed
Warning

Synonyms: (10-(4,5-Dimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)decyl)triphenylphosphonium bromide, Visomitin

A723646-100MG A723646-250MG A723646-50MG
77622-564EA 349.17 USD
77622-564 77622-566 77622-568
SKQ1 Bromide ≥98%
SKQ1 Bromide

Precaución: for Research Use Only

Formula: C₃₆H₄₂BrO₂P
MW: 617.61 g/mol
MDL Number: MFCD29477502
CAS Number: 934826-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR