5,6-Dimethoxy-2H-furo[2,3-h]chromen-2-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Pimpinellin

A723402-10MG A723402-1MG A723402-5MG
77614-228EA 61.8 USD
77614-228 77614-230 77614-232
5,6-Dimethoxy-2H-furo[2,3-h]chromen-2-one ≥98%
5,6-Dimethoxy-2H-furo[2,3-h]chromen-2-one

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₅
MW: 246.22 g/mol
MDL Number: MFCD00597237
CAS Number: 131-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR