Coumarin 343 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,3,6,7-Tetrahydro-11-oxo-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizine-10-carboxylic Acid, 11-oxo-2,3,5,6,7,11-Hexahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinoline-10-carboxylic acid

A723394-100MG A723394-250MG A723394-5G
77602-858EA 46.13 USD
77602-858 77880-485 77880-486
Coumarin 343 ≥98%
Coumarin 343

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₄
MDL Number: MFCD00051335
CAS Number: 55804-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR