Salinomycin ≥98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((2R,5S,6R)-6-((2S,3S,4S,6R)-6-((2S,5S,7R,9S,10S,12R,15R)-2-((2R,5R,6S)-5-Ethyl-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)-15-hydroxy-2,10,12-trimethyl-1,6,8-trioxadispiro[4.1.57.35]pentadec-13-en-9-yl)-3-hydroxy-4-methyl-5-oxooctan-2-yl)-5-methyltetrahydro-2H-pyran-2-yl)butanoic acid

A721014-10MG A721014-1MG
77602-852EA 141.45 USD
77602-852 77669-944
Salinomycin ≥98%
Salinomycin

Precaución: for Research Use Only

Formula: C₄₂H₇₀O₁₁
MW: 751.00 g/mol
MDL Number: MFCD25541652
CAS Number: 53003-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR