(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-((4-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl)amino)benzyl)oxy)-5-oxopentanoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Glu(ODmab)-OH

A720640-1G A720640-100MG A720640-250MG
77614-220EA 212.18 USD
77614-220 77673-976 77673-978
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-((4-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl)amino)benzyl)oxy)-5-oxopentanoic acid ≥98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-((4-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl)amino)benzyl)oxy)-5-oxopentanoic acid

Precaución: for Research Use Only

Formula: C₄₀H₄₄N₂O₈
MDL Number: MFCD00797873
CAS Number: 268730-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR