N-Benzyl-N-((4,5-dihydro-1H-imidazol-2-yl)methyl)aniline ≥98%

Supplier: Ambeed
Warning

Synonyms: Antazoline

A720616-10MG A720616-100MG A720616-1G A720616-250MG A720616-50MG
77627-468EA 39.14 USD
77627-468 77878-561 77878-562 77878-563 77878-564
N-Benzyl-N-((4,5-dihydro-1H-imidazol-2-yl)methyl)aniline ≥98%
N-Benzyl-N-((4,5-dihydro-1H-imidazol-2-yl)methyl)aniline

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃
MW: 265.35 g/mol
MDL Number: MFCD00047013
CAS Number: 91-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR