5-Bromo-2-(trifluoromethoxy)benzene-1-sulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A720338-100MG A720338-1G A720338-250MG
77893-437EA 57 USD
77893-437 77893-438 77893-439
5-Bromo-2-(trifluoromethoxy)benzene-1-sulfonyl chloride ≥95%
5-Bromo-2-(trifluoromethoxy)benzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O₃S
MDL Number: MFCD27932601
CAS Number: 915763-79-0
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR