2,5-Dioxopyrrolidin-1-yl pent-4-ynoate ≥98%

Supplier: Ambeed
Warning

Synonyms: Propargyl-C1-NHS ester

A720327-100MG A720327-1G A720327-250MG
77573-324EA 76 USD
77573-324 77573-326 77686-717
2,5-Dioxopyrrolidin-1-yl pent-4-ynoate ≥98%
2,5-Dioxopyrrolidin-1-yl pent-4-ynoate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.17 g/mol
MDL Number: MFCD22577791
CAS Number: 132178-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR