3,6,9,12,15,18,21,24-Octaoxahexacosane-1,26-diyl bis(4-methylbenzenesulfonate) ≥97%

Supplier: Ambeed
Warning

A720092-250MG A720092-100MG A720092-1G
77598-772EA 104.55 USD
77598-772 77622-554 77622-556
3,6,9,12,15,18,21,24-Octaoxahexacosane-1,26-diyl bis(4-methylbenzenesulfonate) ≥97%
3,6,9,12,15,18,21,24-Octaoxahexacosane-1,26-diyl bis(4-methylbenzenesulfonate)

Precaución: for Research Use Only

Formula: C₃₂H₅₀O₁₄S₂
MDL Number: MFCD25372003
CAS Number: 109635-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR