1-tert-Butyl 2-methyl 4-hydroxypiperidine-1,2-dicarboxylate ≥98%
Supplier: Ambeed
|
Warning
|
A720035-5G
A720035-1G
77614-218EA
302.82
USD
77614-218
77673-974
1-tert-Butyl 2-methyl 4-hydroxypiperidine-1,2-dicarboxylate ≥98%
1-tert-Butyl 2-methyl 4-hydroxypiperidine-1,2-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₂₁NO₅ MW: 259.29 g/mol |
MDL Number:
MFCD08437533 CAS Number: 187753-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...