2-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-(morpholine-4-carbonyl)phenylboronic acid, (2-Fluoro-5-(morpholine-4-carbonyl)phenyl)boronic acid, 2-Fluoro-5-(4-morpholinylcarbonyl)benzeneboronic acid

A719659-100MG A719659-1G A719659-250MG A719659-5G
77884-630EA 55 USD
77884-630 77884-631 77884-632 77884-633
2-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid ≥98%
2-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BFNO₄
MDL Number: MFCD11504847
CAS Number: 1072951-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR