PTP1B inhibitor ≥95%

Supplier: Ambeed
Warning

A719591-1MG A719591-5MG
77669-940EA 47 USD
77669-940 77669-942
PTP1B inhibitor ≥95%
PTP1B inhibitor

Precaución: for Research Use Only

Formula: C₂₆H₁₉Br₂N₃O₇S₃
MDL Number: MFCD09037556
CAS Number: 765317-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR