(4-((1-(tert-Butoxycarbonyl)piperidin-4-yl)oxy)phenyl)boronic acid ≥97%

Supplier: Ambeed
Warning

A717676-100MG A717676-1G
77900-682EA 46 USD
77900-682 77901-314
(4-((1-(tert-Butoxycarbonyl)piperidin-4-yl)oxy)phenyl)boronic acid ≥97%
(4-((1-(tert-Butoxycarbonyl)piperidin-4-yl)oxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₄BNO₅
MDL Number: MFCD12068408
CAS Number: 1224449-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR