tert-Butyl 4-hydroxy-4-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)piperidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A716870-100MG A716870-250MG A716870-1G
77598-766EA 172.2 USD
77598-766 77598-768 77622-548
tert-Butyl 4-hydroxy-4-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)piperidine-1-carboxylate ≥98%
tert-Butyl 4-hydroxy-4-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₃₄BN₃O₅
MW: 407.31 g/mol
MDL Number: MFCD22628001
CAS Number: 877399-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR