8-Ethynyl-1,4-dioxaspiro[4.5]decane ≥98%

Supplier: Ambeed
Warning

A716790-5G A716790-1G
77595-122EA 231.65 USD
77595-122 77622-546
8-Ethynyl-1,4-dioxaspiro[4.5]decane ≥98%
8-Ethynyl-1,4-dioxaspiro[4.5]decane

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MDL Number: MFCD25972927
CAS Number: 96184-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR