3-Ethynylbicyclo[1.1.1]pentane-1-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A716207-1G A716207-100MG A716207-250MG
77749-461EA 1084 USD
77749-461 77749-460 77749-462
3-Ethynylbicyclo[1.1.1]pentane-1-carboxylic acid ≥98%
3-Ethynylbicyclo[1.1.1]pentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.15 g/mol
MDL Number: MFCD30613412
CAS Number: 2091699-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR