1-((3-Aminopropyl)amino)anthracene-9,10-dione hydrochloride ≥95%

Supplier: Ambeed
Warning

A716151-1G A716151-250MG
77893-429EA 124 USD
77893-429 77893-430
1-((3-Aminopropyl)amino)anthracene-9,10-dione hydrochloride ≥95%
1-((3-Aminopropyl)amino)anthracene-9,10-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₇ClN₂O₂
MDL Number: MFCD29918815
CAS Number: 97404-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR