1-(5-Bromo-3-fluoropyridin-2-yl)ethanone ≥98%

Supplier: Ambeed
Warning

A715832-1G A715832-250MG
77900-976EA 101 USD
77900-976 77901-660
1-(5-Bromo-3-fluoropyridin-2-yl)ethanone ≥98%
1-(5-Bromo-3-fluoropyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
MDL Number: MFCD13689180
CAS Number: 1160936-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR