Bis(4-fluorophenyl)iodonium trifluoromethanesulfonate ≥97% (by HPLC, titration analysis)

Supplier: Ambeed
Warning

Synonyms: Bis(4-fluorophenyl)iodonium triflate, Bis(4-fluorophenyl)iodonium Trifluoromethanesulphonate

A715580-250MG A715580-1G
77783-740EA 9 USD
77783-740 77783-739
Bis(4-fluorophenyl)iodonium trifluoromethanesulfonate ≥97% (by HPLC, titration analysis)
Bis(4-fluorophenyl)iodonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₃H₈F₅IO₃S
MW: 466.16 g/mol
MDL Number: MFCD21608479
CAS Number: 732306-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR