2-Methoxy-4-methylphenylboronic acid pinacol ester 95+%

Supplier: Ambeed
Warning

A715560-250MG A715560-5G
77891-411EA 75 USD
77891-411 77891-412
2-Methoxy-4-methylphenylboronic acid pinacol ester 95+%
2-Methoxy-4-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₃
MDL Number: MFCD18732610
CAS Number: 1326662-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR