(2,2-Difluoroethyl)hydrazine hydrochloride ≥95%

Supplier: Ambeed
Warning

A715272-250MG A715272-100MG
77622-544EA 285.31 USD
77622-544 77783-735
(2,2-Difluoroethyl)hydrazine hydrochloride ≥95%
(2,2-Difluoroethyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₂H₇ClF₂N₂
MDL Number: MFCD27959475
CAS Number: 1504582-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR