2-(Prop-2-ynyloxy)acetic acid ≥95%

Supplier: Ambeed
Danger

Synonyms: 2-(Prop-2-yn-1-yloxy)acetic acid

A715238-1G A715238-5G
77598-764EA 269.58 USD
77598-764 77622-542
2-(Prop-2-ynyloxy)acetic acid ≥95%
2-(Prop-2-ynyloxy)acetic acid

Precaución: for Research Use Only

Formula: C₅H₆O₃
MW: 114.1 g/mol
MDL Number: MFCD10694017
CAS Number: 98021-61-5
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR