tert-Butyl ((2-bromopyridin-4-yl)methyl)carbamate ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A714510-1G A714510-100MG A714510-250MG A714510-5G
77684-634EA 121 USD
77684-634 77891-407 77891-408 77891-409
tert-Butyl ((2-bromopyridin-4-yl)methyl)carbamate ≥97%, Ambeed.Inc
tert-Butyl ((2-bromopyridin-4-yl)methyl)carbamate

Precaución: For research use only.

Formula: C₁₁H₁₅BrN₂O₂
MDL Number: MFCD11505921
CAS Number: 1049025-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR