1-(4-Nitrobenzyl)piperazine ≥98%

Supplier: Ambeed
Warning

A714384-1G A714384-250MG
77891-404EA 64 USD
77891-404 77891-405
1-(4-Nitrobenzyl)piperazine ≥98%
1-(4-Nitrobenzyl)piperazine

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD02223757
CAS Number: 58198-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR