Dodecanedioyl dichloride ≥98%

Supplier: Ambeed
Danger

A713642-1G A713642-5G A713642-25G
77602-820EA 15.38 USD
77602-820 77602-822 77627-458
Dodecanedioyl dichloride ≥98%
Dodecanedioyl dichloride

Precaución: for Research Use Only

Formula: C₁₂H₂₀Cl₂O₂
MW: 267.19 g/mol
MDL Number: MFCD00012243
CAS Number: 4834-98-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR