(R)-1-[(R)-tert-Butylphosphinoyl]-2-[(R)-1-(diphenylphosphino)ethyl]ferrocene ≥98%

Supplier: Ambeed
Warning

Synonyms: (Sp)-1-[(R)-tert-Butylphosphinoyl]-2-[(R)-1-(diphenylphosphino)ethyl]ferrocene

A713379-250MG A713379-1G
77747-416EA 201 USD
77747-416 77747-415
(R)-1-[(R)-tert-Butylphosphinoyl]-2-[(R)-1-(diphenylphosphino)ethyl]ferrocene ≥98%
(R)-1-[(R)-tert-Butylphosphinoyl]-2-[(R)-1-(diphenylphosphino)ethyl]ferrocene

Precaución: for Research Use Only

Formula: C₂₈H₃₂FeOP₂
MW: 502.35 g/mol
MDL Number: MFCD22666409
CAS Number: 1221745-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR