tert-Butyl (trans-4-(cyanomethyl)cyclohexyl)carbamate ≥98%

Supplier: Ambeed
Warning

A713170-100MG A713170-250MG
77686-703EA 36 USD
77686-703 77686-704
tert-Butyl (trans-4-(cyanomethyl)cyclohexyl)carbamate ≥98%
tert-Butyl (trans-4-(cyanomethyl)cyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂O₂
MW: 238.33 g/mol
MDL Number: MFCD29060206
CAS Number: 1313279-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR