4-Chloro-1H-indole-2-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A713117-1G A713117-250MG
77602-818EA 31.78 USD
77602-818 77673-956
4-Chloro-1H-indole-2-carboxylic acid ≥97%
4-Chloro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MDL Number: MFCD02664470
CAS Number: 24621-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR