4-Butoxy-2-methylphenylboronic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: 4-N-Butoxy-2-methylphenylboronic acid

A713068-1G A713068-250MG A713068-5G
77873-117EA 45 USD
77873-117 77873-118 77873-119
4-Butoxy-2-methylphenylboronic acid ≥97%
4-Butoxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₃
MDL Number: MFCD05664325
CAS Number: 845551-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR