4,5-Dichloro-2-(m-tolyl)pyridazin-3(2H)-one ≥98%

Supplier: Ambeed
Danger

A712979-1G A712979-250MG
77669-918EA 47.85 USD
77669-918 77669-920
4,5-Dichloro-2-(m-tolyl)pyridazin-3(2H)-one ≥98%
4,5-Dichloro-2-(m-tolyl)pyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₁H₈Cl₂N₂O
MDL Number: MFCD00814448
CAS Number: 41931-13-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR