Methyl 3-(benzyloxy)propanoate ≥95%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyloxy-propionic acid methyl ester

A712267-1G A712267-5G A712267-250MG
77602-810EA 64.58 USD
77602-810 77602-812 77673-954
Methyl 3-(benzyloxy)propanoate ≥95%
Methyl 3-(benzyloxy)propanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
MDL Number: MFCD08274890
CAS Number: 4126-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR