1-Methyl-1H-1,2,3-triazole-4-carboxylic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A712058-1G A712058-5G
77684-630EA 597 USD
77684-630 77886-649
1-Methyl-1H-1,2,3-triazole-4-carboxylic acid ≥98%, Ambeed.Inc
1-Methyl-1H-1,2,3-triazole-4-carboxylic acid

Precaución: For research use only.

Formula: C₄H₅N₃O₂
MDL Number: MFCD08689473
CAS Number: 16681-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR