(S)-4-((2-(Azetidin-1-yl)-1-(4-chloro-3-(trifluoromethyl)phenyl)ethyl)amino)quinazoline-8-carboxamide 98+%

Supplier: Ambeed
Warning

Synonyms: M2698

A711494-1MG A711494-10MG A711494-5MG
77617-932EA 42.23 USD
77617-932 77749-454 77749-455
(S)-4-((2-(Azetidin-1-yl)-1-(4-chloro-3-(trifluoromethyl)phenyl)ethyl)amino)quinazoline-8-carboxamide 98+%
(S)-4-((2-(Azetidin-1-yl)-1-(4-chloro-3-(trifluoromethyl)phenyl)ethyl)amino)quinazoline-8-carboxamide

Precaución: for Research Use Only

Formula: C₂₁H₁₉ClF₃N₅O
MW: 449.86 g/mol
MDL Number: MFCD31744446
CAS Number: 1379545-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR