(S)-3-(Pyridin-3-yl)propyl 1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: GPI-1046

A711280-10MG A711280-1MG A711280-5MG
77614-188EA 98.88 USD
77614-188 77614-190 77614-192
(S)-3-(Pyridin-3-yl)propyl 1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate ≥98%
(S)-3-(Pyridin-3-yl)propyl 1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₂₈N₂O₄
MW: 360.45 g/mol
MDL Number: MFCD00953146
CAS Number: 186452-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR