tert-Butyl (4R)-4-amino-3,3-difluoropiperidine-1-carboxylate ≥95% (by HPLC)

Supplier: Ambeed
Danger

A711259-50MG A711259-5G
77622-530EA 176.13 USD
77622-530 77622-532
tert-Butyl (4R)-4-amino-3,3-difluoropiperidine-1-carboxylate ≥95% (by HPLC)
tert-Butyl (4R)-4-amino-3,3-difluoropiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈F₂N₂O₂
MW: 236.2589 g/mol
MDL Number: MFCD30386986
CAS Number: 1820679-70-6
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR