Zinc protoporphyrin ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Zinc(II) 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate, Protoporphyrin IX zinc(II), Zinc (II) Protoporphyrin IX
A711140-1MG
A711140-10MG
A711140-5MG
A711140-50MG
77614-184EA
21.63
USD
77614-184
77686-696
77686-697
77901-597
Zinc protoporphyrin ≥98%
Zinc protoporphyrin
Precaución: for Research Use Only
Formula:
C₃₄H₃₂N₄O₄Zn MW: 626.034 g/mol |
MDL Number:
MFCD00011612 CAS Number: 15442-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...