2-(4-Hydroxy-3,5-diisopropylbenzylidene)malononitrile ≥98%

Supplier: Ambeed
Danger

Synonyms: AG 1406

A711008-100MG A711008-250MG
77622-528EA 104.03 USD
77622-528 77783-705
2-(4-Hydroxy-3,5-diisopropylbenzylidene)malononitrile ≥98%
2-(4-Hydroxy-3,5-diisopropylbenzylidene)malononitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₂O
MDL Number: MFCD31620835
CAS Number: 71308-34-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR