Methyl 3-nitro-1H-pyrazole-4-carboxylate ≥97%

Supplier: Ambeed
Warning

A709881-1G A709881-5G A709881-250MG
77747-412EA 292 USD
77747-412 77747-414 77747-413
Methyl 3-nitro-1H-pyrazole-4-carboxylate ≥97%
Methyl 3-nitro-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅N₃O₄
MW: 171.11 g/mol
MDL Number: MFCD28155637
CAS Number: 1798715-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR